Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NID1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002508.2 | NP_002499.2 | 2 | 465 | ||
XM_011544195.2 | XP_011542497.1 | 2 | 462 | |||
GenBank mRNA | AB209448.1 | - | 2 | 362 | ||
AK297234.1 | - | 2 | 486 | |||
BC045606.1 | - | 2 | 375 | 5271590 | ||
M30269.1 | - | 2 | 453 |