Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NINJ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004148.3 | NP_004139.2 | 2 | 179 | ||
XM_011518716.1 | XP_011517018.1 | 3 | 252 | |||
GenBank mRNA | BC000298.1 | - | 2 | 144 | 2820092 | |
BC004440.1 | - | 2 | 135 | 2820092 | ||
BC019336.1 | - | 2 | 134 | 3617778 | ||
BC048212.1 | - | 2 | 179 | 5806415 | ||
BT007164.1 | - | 2 | 108 | |||
U72661.1 | - | 2 | 128 | |||
U91512.1 | - | 2 | 128 |