Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NINJ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004148.3 | NP_004139.2 | 3 | 452 | ||
XM_011518716.1 | XP_011517018.1 | 4 | 525 | |||
GenBank mRNA | AK094530.1 | - | 1 | 1208 | ||
BC000298.1 | - | 3 | 417 | 2820092 | ||
BC004440.1 | - | 3 | 408 | 2820092 | ||
BC019336.1 | - | 3 | 407 | 3617778 | ||
BC048212.1 | - | 3 | 452 | 5806415 | ||
BT007164.1 | - | 3 | 381 | |||
U72661.1 | - | 3 | 401 | |||
U91512.1 | - | 3 | 401 |