Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NKG7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005601.3 | NP_005592.1 | 3 | 590 | ||
XM_005258955.3 | XP_005259012.1 | 3 | 467 | |||
XM_006723228.3 | XP_006723291.1 | 2 | 425 | |||
GenBank mRNA | AI313472.1 | - | 2 | 236 | ||
BC015759.1 | - | 3 | 519 | 4849734 | ||
S69115.1 | - | 3 | 599 |