Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NQO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000904.4 | NP_000895.2 | 4 | 575 | ||
NM_001290221.1 | NP_001277150.1 | 7 | 1090 | |||
NM_001290222.1 | NP_001277151.1 | 4 | 575 | |||
NM_001318940.1 | NP_001305869.1 | 4 | 515 | |||
GenBank mRNA | AK056981.1 | - | 7 | 932 | ||
AK311746.1 | - | 4 | 534 | |||
BC006096.2 | - | 3 | 306 | 3507945 | ||
BI333292.1 | - | 4 | 514 | |||
BI670476.1 | - | 4 | 563 | |||
CD014013.1 | - | 4 | 527 | |||
J02888.1 | - | 4 | 395 |