Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NPHS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004646.3 | NP_004637.1 | 26 | 3482 | ||
GenBank mRNA | AF035835.1 | - | 26 | 3326 | ||
AF126957.1 | - | 4 | 113 | |||
AF190637.1 | - | 26 | 3326 | |||
EU642886.1 | - | 26 | 3326 | |||
EU642887.1 | - | 26 | 3325 |