Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ROR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083592.1 | NP_001077061.1 | 7 | 1440 | ||
NM_005012.3 | NP_005003.2 | 7 | 1440 | |||
XM_011541526.1 | XP_011539828.1 | 7 | 1039 | |||
XM_017001376.1 | XP_016856865.1 | 6 | 1340 | |||
XM_017001377.1 | XP_016856866.1 | 9 | 3983 | |||
GenBank mRNA | AK308088.1 | - | 7 | 1446 | ||
AK309189.1 | - | 4 | 682 | |||
BC080541.1 | - | 7 | 1440 | |||
BC128386.1 | - | 8 | 1050 | |||
M97675.1 | - | 7 | 1416 |