Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ROR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318204.1 | NP_001305133.1 | 6 | 892 | ||
NM_004560.3 | NP_004551.2 | 6 | 892 | |||
XM_005252008.4 | XP_005252065.1 | 6 | 857 | |||
XM_006717121.3 | XP_006717184.1 | 6 | 1514 | |||
XM_017014762.1 | XP_016870251.1 | 6 | 2589 | |||
XM_017014763.1 | XP_016870252.1 | 6 | 768 | |||
GenBank mRNA | AB209154.1 | - | 8 | 1161 | ||
BC033697.1 | - | 6 | 879 | |||
BC051273.1 | - | 6 | 704 | |||
BC130522.1 | - | 6 | 780 | 40146553 | ||
BI089313.1 | - | 4 | 252 | |||
M97639.1 | - | 6 | 892 |