Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NTS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006183.4 | NP_006174.1 | 1 | 139 | ||
GenBank mRNA | AF157682.1 | - | 1 | 115 | ||
AK312066.1 | - | 1 | 139 | |||
BC010918.1 | - | 1 | 141 | 4285844 | ||
CR541936.1 | - | 1 | 30 | |||
U91618.1 | - | 1 | 58 |