Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NVL SIRNA Assays ›
Gene Symbol:
NVL
Gene Name:
nuclear VCP-like
Gene Aliases:
NVL2
Chromosome Location:
Chr.1: 224227334 - 224330201 on Build GRCh38
UniGene ID:
Hs.497867
Species:
Human
siRNA ID
HSS107374
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NVL
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001243146.1
NP_001230075.1
10, 11
1064
NM_001243147.1
NP_001230076.1
11, 12
1115
NM_002533.3
NP_002524.2
12, 13
1388
NM_206840.2
NP_996671.1
11, 12
1314
XM_011544196.1
XP_011542498.1
13, 14
1452
XM_011544198.1
XP_011542500.1
12, 13
1407
XM_011544199.1
XP_011542501.1
12, 13
1361
XM_011544200.1
XP_011542502.1
11, 12
1335
XM_017001378.1
XP_016856867.1
14, 15
1561
XM_017001379.1
XP_016856868.1
13, 14
1480
XM_017001380.1
XP_016856869.1
14, 15
1561
XM_017001381.1
XP_016856870.1
14, 15
1558
XM_017001382.1
XP_016856871.1
13, 14
1491
XM_017001383.1
XP_016856872.1
12, 13
1410
XM_017001384.1
XP_016856873.1
14, 15
1561
XM_017001385.1
XP_016856874.1
14, 15
1561
XM_017001386.1
XP_016856875.1
10, 11
1254
GenBank mRNA
AK297070.1
-
12, 13
1347
AK297396.1
-
10, 11
1015
AK298244.1
-
11, 12
1064
AK298826.1
-
11, 12
1266
BC012105.1
-
10, 11
975
4636004
U68140.1
-
12, 13
1382
U78772.1
-
11, 12
1295
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
ribosome biogenesis
positive regulation of telomerase activity
protein binding
ATP binding
poly(A) RNA binding
preribosome binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wzvj9:80/10.239.161.253:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0