Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NVL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243146.1 | NP_001230075.1 | 12 | 1455 | ||
NM_001243147.1 | NP_001230076.1 | 13 | 1506 | |||
NM_002533.3 | NP_002524.2 | 14 | 1779 | |||
NM_206840.2 | NP_996671.1 | 13 | 1705 | |||
XM_011544196.1 | XP_011542498.1 | 15 | 1843 | |||
XM_011544198.1 | XP_011542500.1 | 14 | 1798 | |||
XM_011544199.1 | XP_011542501.1 | 14 | 1752 | |||
XM_011544200.1 | XP_011542502.1 | 13 | 1726 | |||
XM_017001378.1 | XP_016856867.1 | 16 | 1952 | |||
XM_017001379.1 | XP_016856868.1 | 15 | 1871 | |||
XM_017001380.1 | XP_016856869.1 | 16 | 1952 | |||
XM_017001381.1 | XP_016856870.1 | 16 | 1949 | |||
XM_017001382.1 | XP_016856871.1 | 15 | 1882 | |||
XM_017001383.1 | XP_016856872.1 | 14 | 1801 | |||
XM_017001384.1 | XP_016856873.1 | 16 | 1952 | |||
XM_017001385.1 | XP_016856874.1 | 16 | 1952 | |||
XM_017001386.1 | XP_016856875.1 | 12 | 1645 | |||
GenBank mRNA | AK297070.1 | - | 14 | 1738 | ||
AK297396.1 | - | 12 | 1406 | |||
AK298244.1 | - | 13 | 1455 | |||
AK298826.1 | - | 13 | 1657 | |||
BC012105.1 | - | 12 | 1366 | 4636004 | ||
U68140.1 | - | 14 | 1773 | |||
U78772.1 | - | 13 | 1686 |