Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OAS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001032409.2 | NP_001027581.1 | 5 | 1246 | ||
NM_001320151.1 | NP_001307080.1 | 5 | 1246 | |||
NM_002534.3 | NP_002525.2 | 5 | 1246 | |||
NM_016816.3 | NP_058132.2 | 5 | 1246 | |||
XM_006719434.1 | XP_006719497.1 | 5 | 1073 | |||
XM_011538413.1 | XP_011536715.1 | 5 | 1049 | |||
XM_017019361.1 | XP_016874850.1 | 5 | 1049 | |||
XM_017019362.1 | XP_016874851.1 | 5 | 898 | |||
GenBank mRNA | AA085948.1 | - | 2 | 365 | ||
AJ629455.1 | - | 5 | 1016 | |||
AK123528.1 | - | 1 | 2095 | |||
AK223006.1 | - | 5 | 1005 | |||
AK225384.1 | - | 5 | 1046 | |||
AK291003.1 | - | 5 | 1050 | |||
AY730627.1 | - | 5 | 992 | |||
AY730628.1 | - | 5 | 992 | |||
BC000562.2 | - | 5 | 1053 | 3162421 | ||
BC061587.1 | - | 5 | 1014 | 6502031 | ||
BC071981.1 | - | 5 | 1049 | 4053409 | ||
BE048946.1 | - | 1 | 378 | |||
BT006785.1 | - | 5 | 983 | |||
D00068.1 | - | 5 | 1046 | |||
X02661.1 | - | 2 | 293 | |||
X02874.1 | - | 5 | 1016 | |||
X02875.1 | - | 2 | 116 | |||
X04371.1 | - | 5 | 1076 |