Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OAS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006187.3 | NP_006178.2 | 6 | 1316 | ||
XM_005253889.3 | XP_005253946.1 | 6 | 1237 | |||
XM_017019363.1 | XP_016874852.1 | 6 | 1237 | |||
GenBank mRNA | AB044545.1 | - | 6 | 1397 | ||
AF063613.1 | - | 6 | 1252 | |||
AK292949.1 | - | 6 | 1286 | |||
BC113746.1 | - | 6 | 1291 | 40036464 | ||
BM474470.1 | - | 1 | 14 |