Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ODC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287188.1 | NP_001274117.1 | 11 | 1544 | ||
NM_001287189.1 | NP_001274118.1 | 11 | 1536 | |||
NM_001287190.1 | NP_001274119.1 | 11 | 1382 | |||
NM_002539.2 | NP_002530.1 | 11 | 1644 | |||
GenBank mRNA | AK292352.1 | - | 11 | 1517 | ||
AK301956.1 | - | 11 | 1317 | |||
AK312766.1 | - | 9 | 1084 | |||
BC025296.1 | - | 11 | 1384 | 5088190 | ||
M16650.2 | - | 10 | 1170 | |||
M20372.1 | - | 7 | 751 | |||
X55362.1 | - | 11 | 1385 |