Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OGN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014057.4 | NP_054776.1 | 3 | 470 | ||
NM_024416.4 | NP_077727.3 | 3 | 389 | |||
NM_033014.3 | NP_148935.1 | 3 | 752 | |||
GenBank mRNA | AF086912.1 | - | 3 | 317 | ||
AF100758.1 | - | 3 | 317 | |||
AF202167.1 | - | 2 | 242 | |||
AK075521.1 | - | 2 | 281 | |||
AK129845.1 | - | 2 | 274 | |||
AK289628.1 | - | 3 | 467 | |||
AK295433.1 | - | 3 | 389 | |||
AL110267.1 | - | 2 | 259 | |||
BC037273.2 | - | 2 | 258 | 5265880 | ||
BC095443.1 | - | 2 | 274 | 30405707 | ||
BG772245.1 | - | 3 | 392 | |||
BX646984.1 | - | 3 | 427 | |||
CR533516.1 | - | 2 | 191 |