Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OGN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014057.4 | NP_054776.1 | 6 | 985 | ||
NM_024416.4 | NP_077727.3 | 6 | 904 | |||
NM_033014.3 | NP_148935.1 | 6 | 1267 | |||
GenBank mRNA | AF086912.1 | - | 6 | 832 | ||
AF088023.1 | - | 3 | 323 | |||
AF100758.1 | - | 6 | 832 | |||
AF202167.1 | - | 5 | 757 | |||
AK075521.1 | - | 5 | 796 | |||
AK129845.1 | - | 5 | 789 | |||
AK289628.1 | - | 6 | 982 | |||
AK295433.1 | - | 6 | 904 | |||
AL110267.1 | - | 5 | 774 | |||
AL133114.1 | - | 2 | 250 | |||
BC037273.2 | - | 5 | 773 | 5265880 | ||
BC095443.1 | - | 5 | 789 | 30405707 | ||
CR533516.1 | - | 5 | 706 |