Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OLR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172632.1 | NP_001166103.1 | 5 | 698 | ||
NM_001172633.1 | NP_001166104.1 | 5 | 722 | |||
NM_002543.3 | NP_002534.1 | 6 | 838 | |||
GenBank mRNA | AB010710.1 | - | 6 | 785 | ||
AB102861.1 | - | 6 | 724 | |||
AF035776.1 | - | 6 | 770 | |||
AK292124.1 | - | 6 | 779 | |||
AK295409.1 | - | 5 | 639 | |||
AK298040.1 | - | 6 | 648 | |||
AK316353.1 | - | 4 | 428 | |||
AL551100.3 | - | 5 | 646 | |||
AX429248.1 | - | 6 | 785 | |||
BC022295.1 | - | 6 | 779 | 4722086 | ||
CB955704.1 | - | 2 | 262 |