Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OPRD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000911.3 | NP_000902.3 | 2 | 619 | ||
GenBank mRNA | EF057065.1 | - | 2 | 377 | ||
EU883570.1 | - | 2 | 377 | |||
U07882.2 | - | 2 | 610 | |||
U10504.1 | - | 2 | 377 |
Set Membership: |
G-protein coupled receptor |