Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ORC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006190.4 | NP_006181.1 | 10, 11 | 1072 | ||
NR_033915.1 | 10, 11 | 1072 | ||||
XM_006712555.3 | XP_006712618.1 | 10, 11 | 1019 | |||
XM_011511252.2 | XP_011509554.1 | 11, 12 | 1134 | |||
XM_011511253.2 | XP_011509555.1 | 11, 12 | 1006 | |||
GenBank mRNA | AK302614.1 | - | 10, 11 | 1044 | ||
AK316433.1 | - | 10, 11 | 973 | |||
BC014834.1 | - | 10, 11 | 1015 | 2899929 | ||
U27459.1 | - | 10, 11 | 1006 | |||
U40268.2 | - | 10, 11 | 987 |