Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLDN11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001185056.1 | NP_001171985.1 | 3 | 327 | ||
NM_005602.5 | NP_005593.2 | 3 | 657 | |||
GenBank mRNA | AJ245901.1 | - | 3 | 455 | ||
AK294087.1 | - | 3 | 298 | |||
AK312923.1 | - | 3 | 654 | |||
BC013577.1 | - | 3 | 645 | 3895040 | ||
BT019402.1 | - | 3 | 455 | |||
BT019403.1 | - | 3 | 455 | |||
DA258090.1 | - | 3 | 327 |