Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P2RX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256796.1 | NP_001243725.1 | 10, 11 | 1336 | ||
NM_001261397.1 | NP_001248326.1 | 9, 10 | 1207 | |||
NM_002560.2 | NP_002551.2 | 9, 10 | 1288 | |||
NR_046372.1 | 10, 11 | 1326 | ||||
NR_046373.1 | 8, 9 | 1140 | ||||
XM_011538416.2 | XP_011536718.1 | 5, 6 | 651 | |||
GenBank mRNA | AF000234.1 | - | 9, 10 | 884 | ||
AK293599.1 | - | 6, 7 | 778 | |||
AK293660.1 | - | 8, 9 | 923 | |||
AK297704.1 | - | 8, 9 | 872 | |||
AK301080.1 | - | 9, 10 | 972 | |||
AK308549.1 | - | 10, 11 | 1058 | |||
AK308868.1 | - | 9, 10 | 1979 | |||
BC017458.1 | - | 9, 10 | 1017 | |||
BC033826.1 | - | 9, 10 | 1008 | 5216449 | ||
BT019738.1 | - | 9, 10 | 984 | |||
BT019739.1 | - | 9, 10 | 903 | |||
JF807485.1 | - | 2, 3 | 141 | |||
JX471561.1 | - | 2, 3 | 180 | |||
U83993.1 | - | 9, 10 | 1293 | |||
Y07684.1 | - | 9, 10 | 1011 |