Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P2RX7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002562.5 | NP_002553.3 | 9 | 1058 | ||
NR_033948.1 | 10 | 1197 | ||||
NR_033949.1 | 10 | 1197 | ||||
NR_033950.1 | 8 | 1074 | ||||
NR_033951.1 | 9 | 1058 | ||||
NR_033952.1 | 8 | 985 | ||||
NR_033953.1 | 8 | 898 | ||||
NR_033954.1 | 8 | 961 | ||||
NR_033955.1 | 8 | 921 | ||||
NR_033956.1 | 7 | 848 | ||||
XM_011538419.2 | XP_011536721.1 | 8 | 898 | |||
XM_011538420.2 | XP_011536722.1 | 7 | 565 | |||
XM_017019364.1 | XP_016874853.1 | 9 | 1211 | |||
XM_017019365.1 | XP_016874854.1 | 8 | 939 | |||
XM_017019366.1 | XP_016874855.1 | 8 | 926 | |||
XM_017019367.1 | XP_016874856.1 | 7 | 764 | |||
GenBank mRNA | AB209709.1 | - | 9 | 979 | ||
AK090866.1 | - | 8 | 775 | |||
AK225163.1 | - | 9 | 944 | |||
AK290405.1 | - | 9 | 1006 | |||
AK296432.1 | - | 9 | 1006 | |||
AY847298.1 | - | 9 | 915 | |||
AY847299.1 | - | 8 | 842 | |||
AY847300.1 | - | 8 | 818 | |||
AY847301.1 | - | 7 | 648 | |||
AY847302.1 | - | 7 | 705 | |||
AY847303.1 | - | 10 | 1054 | |||
AY847304.1 | - | 10 | 1054 | |||
BC007679.2 | - | 8 | 859 | 3628076 | ||
BC011913.2 | - | 9 | 984 | 4298811 | ||
BC121157.1 | - | 9 | 984 | |||
BC121158.1 | - | 8 | 1000 | |||
DQ399293.1 | - | 8 | 778 | |||
GQ180122.1 | - | 9 | 924 | |||
Y09561.1 | - | 9 | 941 |