Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PABPC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030979.2 | NP_112241.2 | 1 | 330 | ||
GenBank mRNA | AF132026.1 | - | 1 | 292 | ||
AK313336.1 | - | 1 | 312 | |||
AL136900.1 | - | 1 | 313 | |||
BC027617.2 | - | 1 | 296 | 4829121 | ||
BC045608.1 | - | 1 | 306 | 5273503 | ||
BG718483.1 | - | 1 | 335 |