Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAX9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006194.3 | NP_006185.1 | 5 | 1612 | ||
GenBank mRNA | AB248958.1 | - | 4 | 1055 | ||
AK310339.1 | - | 5 | 497 | |||
AK313627.1 | - | 4 | 1067 | |||
BC001159.1 | - | 4 | 1075 | 3534582 | ||
U59628.1 | - | 3 | 746 | |||
X92850.1 | - | 3 | 862 |