Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PCM1 SIRNA Assays ›
Gene Symbol:
PCM1
Gene Name:
pericentriolar material 1
Gene Aliases:
PTC4, RET/PCM-1
Chromosome Location:
Chr.8: 17922763 - 18029948 on Build GRCh38
UniGene ID:
Hs.491148
Species:
Human
siRNA ID
HSS107659
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PCM1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001315507.1
NP_001302436.1
13
2325
NM_001315508.1
NP_001302437.1
13
2328
NM_006197.3
NP_006188.3
13
2325
XM_006716336.3
XP_006716399.1
15
2707
XM_006716340.3
XP_006716403.1
15
2707
XM_006716349.3
XP_006716412.1
13
2401
XM_011544528.2
XP_011542830.1
15
2707
XM_011544529.2
XP_011542831.1
17
3013
XM_011544530.2
XP_011542832.1
16
2829
XM_011544532.2
XP_011542834.1
15
2704
XM_011544533.2
XP_011542835.1
15
2707
XM_011544534.2
XP_011542836.1
15
2707
XM_011544535.2
XP_011542837.1
15
2707
XM_011544540.2
XP_011542842.1
15
2635
XM_017013472.1
XP_016868961.1
16
2823
XM_017013473.1
XP_016868962.1
16
2819
XM_017013474.1
XP_016868963.1
14
2515
XM_017013475.1
XP_016868964.1
16
2651
XM_017013476.1
XP_016868965.1
13
2235
XM_017013477.1
XP_016868966.1
16
2819
XM_017013478.1
XP_016868967.1
16
2818
XM_017013479.1
XP_016868968.1
15
2705
XM_017013480.1
XP_016868969.1
13
2398
XM_017013481.1
XP_016868970.1
15
2701
XM_017013482.1
XP_016868971.1
13
2419
XM_017013483.1
XP_016868972.1
15
2700
XM_017013484.1
XP_016868973.1
12
2126
XM_017013485.1
XP_016868974.1
15
2704
XM_017013486.1
XP_016868975.1
14
2515
XM_017013487.1
XP_016868976.1
14
2518
XM_017013488.1
XP_016868977.1
14
2515
XM_017013489.1
XP_016868978.1
16
2818
XM_017013490.1
XP_016868979.1
16
2818
XM_017013491.1
XP_016868980.1
13
2401
XM_017013492.1
XP_016868981.1
13
2398
XM_017013493.1
XP_016868982.1
15
2701
XM_017013494.1
XP_016868983.1
13
2398
XM_017013495.1
XP_016868984.1
15
2538
XM_017013496.1
XP_016868985.1
12
2121
XM_017013497.1
XP_016868986.1
13
2398
XM_017013498.1
XP_016868987.1
13
2401
XM_017013499.1
XP_016868988.1
15
2541
XM_017013500.1
XP_016868989.1
13
2398
XM_017013501.1
XP_016868990.1
15
2700
XM_017013502.1
XP_016868991.1
12
2121
XM_017013503.1
XP_016868992.1
14
2515
XM_017013504.1
XP_016868993.1
13
2419
XM_017013505.1
XP_016868994.1
15
2538
XM_017013506.1
XP_016868995.1
13
2398
XM_017013507.1
XP_016868996.1
15
2702
XM_017013508.1
XP_016868997.1
14
2342
GenBank mRNA
AK307583.1
-
14
2309
AL712077.1
-
4
496
AX785390.1
-
11
1906
BC133052.1
-
12
1978
8381608
BC140946.1
-
13
2154
9056758
L27841.1
-
13
2315
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
G2/M transition of mitotic cell cycle
neuron migration
interkinetic nuclear migration
cytoplasmic microtubule organization
microtubule anchoring
microtubule anchoring at centrosome
nonmotile primary cilium assembly
intraciliary transport involved in cilium morphogenesis
cilium assembly
negative regulation of neurogenesis
centrosome organization
protein localization to centrosome
positive regulation of intracellular protein transport
neuronal stem cell population maintenance
protein binding
identical protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-jlg4l:80/100.66.77.222:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline