Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCNT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006031.5 | NP_006022.3 | 38, 39 | 8856 | ||
XM_005261124.4 | XP_005261181.1 | 39, 40 | 8908 | |||
XM_011529594.2 | XP_011527896.1 | 39, 40 | 8956 | |||
XM_017028363.1 | XP_016883852.1 | 39, 40 | 9077 | |||
GenBank mRNA | AF515282.1 | - | 38, 39 | 8749 | ||
U52962.1 | - | 39, 40 | 8768 |