Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCNT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001315529.1 | NP_001302458.1 | 41 | 9177 | ||
NM_006031.5 | NP_006022.3 | 41 | 9252 | |||
XM_005261124.4 | XP_005261181.1 | 42 | 9304 | |||
XM_011529594.2 | XP_011527896.1 | 42 | 9352 | |||
XM_017028362.1 | XP_016883851.1 | 41 | 9034 | |||
XM_017028363.1 | XP_016883852.1 | 42 | 9473 | |||
GenBank mRNA | AB007862.3 | - | 41 | 9178 | ||
AF515282.1 | - | 41 | 9145 | |||
U52962.1 | - | 42 | 9164 |