Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCSK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000439.4 | NP_000430.3 | 14 | 2253 | ||
NM_001177875.1 | NP_001171346.1 | 14 | 1911 | |||
GenBank mRNA | AB208874.1 | - | 14 | 1929 | ||
AK303888.1 | - | 14 | 1911 | |||
AK308575.1 | - | 8 | 1159 | |||
BC130295.1 | - | 14 | 2135 | 40146326 | ||
BC136486.1 | - | 14 | 2135 | 9020475 | ||
M90753.1 | - | 14 | 2223 | |||
X64810.1 | - | 14 | 2203 |