Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCYT1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001312673.1 | NP_001299602.1 | 5 | 533 | ||
NM_005017.3 | NP_005008.2 | 6 | 597 | |||
GenBank mRNA | AK304534.1 | - | 5 | 390 | ||
AK309073.1 | - | 4 | 1221 | |||
AK309300.1 | - | 4 | 1226 | |||
AK313450.1 | - | 5 | 455 | |||
BC046355.1 | - | 6 | 558 | 5744747 | ||
L28957.1 | - | 5 | 430 |