Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCYT1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001312673.1 | NP_001299602.1 | 5, 6 | 628 | ||
NM_005017.3 | NP_005008.2 | 6, 7 | 692 | |||
GenBank mRNA | AK304534.1 | - | 5, 6 | 485 | ||
AK309073.1 | - | 4, 5 | 1316 | |||
AK309300.1 | - | 4, 5 | 1321 | |||
BC046355.1 | - | 6, 7 | 653 | 5744747 | ||
L28957.1 | - | 5, 6 | 525 |