Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDE4C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000923.4 | NP_000914.2 | 4 | 690 | ||
NM_001098818.2 | NP_001092288.1 | 3 | 415 | |||
NM_001098819.2 | NP_001092289.1 | 3 | 468 | |||
NR_040546.1 | 3 | 415 | ||||
XM_011528056.2 | XP_011526358.1 | 3 | 421 | |||
XM_017026866.1 | XP_016882355.1 | 4 | 699 | |||
GenBank mRNA | AK295050.1 | - | 3 | 931 | ||
AK300888.1 | - | 3 | 365 | |||
AK300983.1 | - | 3 | 378 | |||
AY429558.1 | - | 3 | 340 | |||
U66346.1 | - | 4 | 690 | |||
U66347.1 | - | 3 | 645 | |||
U88712.1 | - | 3 | 468 | |||
U88713.1 | - | 3 | 415 | |||
Z46632.1 | - | 3 | 562 |