Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PDE4D SIRNA Assays ›
Gene Symbol:
PDE4D
Gene Name:
phosphodiesterase 4D
Gene Aliases:
ACRDYS2, DPDE3, HSPDE4D, PDE43, PDE4DN2, STRK1
Chromosome Location:
Chr.5: 58969038 - 60488098 on Build GRCh38
UniGene ID:
Hs.117545
Species:
Human
siRNA ID
HSS107724
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PDE4D
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001104631.1
NP_001098101.1
14
2163
NM_001165899.1
NP_001159371.1
16
2054
NM_001197218.1
NP_001184147.1
14
1902
NM_001197219.1
NP_001184148.1
14
1680
NM_001197220.1
NP_001184149.1
14
2318
NM_001197221.1
NP_001184150.1
10
1638
NM_001197222.1
NP_001184151.1
9
1724
NM_001197223.1
NP_001184152.1
9
1514
NM_006203.4
NP_006194.2
14
1706
XM_005248537.3
XP_005248594.1
14
2860
XM_005248538.4
XP_005248595.1
15
4761
XM_011543469.1
XP_011541771.1
17
2143
XM_011543470.2
XP_011541772.1
17
2067
XM_011543471.1
XP_011541773.1
16
2020
XM_011543473.1
XP_011541775.1
16
1968
XM_011543477.2
XP_011541779.1
16
1970
XM_017009565.1
XP_016865054.1
17
2180
XM_017009566.1
XP_016865055.1
17
2070
XM_017009567.1
XP_016865056.1
15
1808
XM_017009568.1
XP_016865057.1
17
2054
XM_017009569.1
XP_016865058.1
16
1933
XM_017009570.1
XP_016865059.1
13
2753
XM_017009571.1
XP_016865060.1
16
1976
GenBank mRNA
AF012073.1
-
14
1916
AF012074.1
-
10
1652
AF536975.1
-
9
1321
AF536976.1
-
16
1877
AF536977.1
-
14
1680
AJ250852.1
-
10
1248
AJ250854.1
-
15
1843
AY245866.1
-
16
2019
AY245867.1
-
14
2318
AY388960.1
-
14
1601
BC036319.1
-
9
1516
4828512
L20970.1
-
14
1714
U02882.1
-
13
1484
U50157.1
-
9
1322
U50158.1
-
10
1236
U50159.1
-
14
1657
Back To Top
More Information
Panther Classification:
Molecular Function -
phosphodiesterase
Gene Ontology Categories:
Function(s)
Process(es)
regulation of heart rate
cAMP catabolic process
smooth muscle contraction
aging
regulation of receptor activity
regulation of release of sequestered calcium ion into cytosol by sarcoplasmic reticulum
cAMP-mediated signaling
neutrophil chemotaxis
regulation of cAMP metabolic process
positive regulation of interferon-gamma production
positive regulation of interleukin-2 production
positive regulation of interleukin-5 production
negative regulation of peptidyl-serine phosphorylation
multicellular organism growth
negative regulation of heart contraction
T cell receptor signaling pathway
regulation of ryanodine-sensitive calcium-release channel activity
establishment of endothelial barrier
cellular response to lipopolysaccharide
cellular response to epinephrine stimulus
adrenergic receptor signaling pathway
regulation of cardiac muscle cell contraction
adrenergic receptor signaling pathway involved in positive regulation of heart rate
regulation of cell communication by electrical coupling involved in cardiac conduction
negative regulation of relaxation of cardiac muscle
3',5'-cyclic-nucleotide phosphodiesterase activity
3',5'-cyclic-AMP phosphodiesterase activity
protein binding
drug binding
enzyme binding
cAMP binding
beta-2 adrenergic receptor binding
ion channel binding
metal ion binding
ATPase binding
scaffold protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-4vtwv:80/10.239.167.181:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline