Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDE8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243137.1 | NP_001230066.1 | 14, 15 | 1336 | ||
NM_002605.2 | NP_002596.1 | 14, 15 | 1538 | |||
NM_173454.1 | NP_775656.1 | 13, 14 | 1400 | |||
XM_011521668.2 | XP_011519970.1 | 7, 8 | 1672 | |||
XM_017022308.1 | XP_016877797.1 | 11, 12 | 1769 | |||
XM_017022309.1 | XP_016877798.1 | 12, 13 | 1136 | |||
XM_017022310.1 | XP_016877799.1 | 14, 15 | 2075 | |||
XM_017022311.1 | XP_016877800.1 | 15, 16 | 2273 | |||
GenBank mRNA | AF056490.1 | - | 12, 13 | 1001 | ||
AF332653.1 | - | 14, 15 | 1485 | |||
AF388183.1 | - | 14, 15 | 1531 | |||
AF388184.1 | - | 13, 14 | 1393 | |||
AF388185.1 | - | 12, 13 | 1314 | |||
AF388186.1 | - | 13, 14 | 1452 | |||
AF388187.1 | - | 15, 16 | 1729 | |||
AK074280.1 | - | 14, 15 | 1336 | |||
AK127232.1 | - | 11, 12 | 2819 | |||
BC060762.1 | - | 15, 16 | 1601 | 30344110 | ||
BC075822.1 | - | 14, 15 | 1429 | 6296489 |