Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PDE9A SIRNA Assays ›
Gene Symbol:
PDE9A
Gene Name:
phosphodiesterase 9A
Gene Aliases:
HSPDE9A2
Chromosome Location:
Chr.21: 42653752 - 42775509 on Build GRCh38
UniGene ID:
Hs.473927
Species:
Human
siRNA ID
HSS107745
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PDE9A
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001001567.1
NP_001001567.1
12
905
NM_001001568.1
NP_001001568.1
10
756
NM_001001569.1
NP_001001569.1
9
701
NM_001001570.1
NP_001001570.1
12
926
NM_001001571.1
NP_001001571.1
11
834
NM_001001572.1
NP_001001572.1
9
712
NM_001001573.1
NP_001001573.1
10
892
NM_001001574.1
NP_001001574.1
11
827
NM_001001575.1
NP_001001575.1
8
657
NM_001001576.1
NP_001001576.1
8
616
NM_001001577.1
NP_001001577.1
9
735
NM_001001578.1
NP_001001578.1
10
779
NM_001001579.1
NP_001001579.1
10
772
NM_001001580.1
NP_001001580.1
7
572
NM_001001581.1
NP_001001581.1
11
936
NM_001001582.1
NP_001001582.1
12
1014
NM_001001583.1
NP_001001583.1
12
1007
NM_001001584.2
NP_001001584.1
11
835
NM_001001585.1
NP_001001585.1
11
959
NM_001315533.1
NP_001302462.1
13
981
NM_002606.2
NP_002597.1
13
1085
XM_011529598.2
XP_011527900.1
13
1007
XM_011529600.2
XP_011527902.1
13
1058
XM_017028366.1
XP_016883855.1
11
823
GenBank mRNA
AF048837.1
-
13
1082
AF067223.1
-
13
1034
AF067224.1
-
12
853
AF067225.1
-
10
695
AF067226.1
-
9
640
AK001165.1
-
10
687
AK001307.1
-
12
876
AK127770.1
-
3
2436
AK294112.1
-
10
722
AK308753.1
-
7
561
AK314679.1
-
13
1058
AY196299.1
-
12
899
AY196300.1
-
11
807
AY196301.1
-
9
685
AY196302.1
-
10
865
AY196303.1
-
11
800
AY196304.1
-
8
630
AY196305.1
-
8
589
AY196306.1
-
9
708
AY196307.1
-
10
752
AY196308.1
-
10
745
AY196309.1
-
7
545
AY196310.1
-
11
909
AY196311.1
-
12
987
AY196312.1
-
12
980
AY196313.1
-
11
807
AY196314.1
-
11
932
AY242121.1
-
11
771
AY701187.1
-
13
978
BC009047.1
-
12
879
3874635
BT007016.1
-
12
842
KU178246.1
-
12
842
KU178247.1
-
13
1022
KU178248.1
-
9
638
Back To Top
More Information
Panther Classification:
Molecular Function -
phosphodiesterase
Gene Ontology Categories:
Function(s)
Process(es)
signal transduction
positive regulation of cardiac muscle hypertrophy
cGMP-mediated signaling
cGMP metabolic process
cGMP catabolic process
3',5'-cyclic-nucleotide phosphodiesterase activity
protein binding
metal ion binding
3',5'-cyclic-GMP phosphodiesterase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-6lbnp:80/10.239.161.162:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline