Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDGFRA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006206.4 | NP_006197.1 | 4 | 853 | ||
XM_005265743.1 | XP_005265800.1 | 4 | 637 | |||
XM_006714039.3 | XP_006714102.1 | 5 | 898 | |||
XM_006714041.3 | XP_006714104.1 | 5 | 901 | |||
XM_011534385.2 | XP_011532687.1 | 4 | 891 | |||
XM_017008280.1 | XP_016863769.1 | 4 | 1068 | |||
XM_017008281.1 | XP_016863770.1 | 4 | 1134 | |||
XM_017008282.1 | XP_016863771.1 | 4 | 782 | |||
GenBank mRNA | AK308353.1 | - | 4 | 683 | ||
AK311006.1 | - | 3 | 340 | |||
AK316578.1 | - | 4 | 655 | |||
BC015186.1 | - | 4 | 670 | |||
BC063414.1 | - | 4 | 637 | 5205969 | ||
M21574.1 | - | 4 | 661 | |||
M22734.1 | - | 650 |