Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199900.1 | NP_001186829.1 | 1 | 217 | ||
NM_002611.4 | NP_002602.2 | 1 | 217 | |||
GenBank mRNA | AK290522.1 | - | 1 | 116 | ||
AK293600.1 | - | 1 | 116 | |||
AK297089.1 | - | 1 | 172 | |||
BC005811.2 | - | 1 | 105 | 3831465 | ||
BC040478.1 | - | 1 | 160 | 4690182 | ||
BC063137.1 | - | 1 | 131 | 4272593 | ||
BQ071206.1 | - | 116 | ||||
L42451.1 | - | 1 | 104 |