Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002612.3 | NP_002603.1 | 10 | 1399 | ||
GenBank mRNA | AK096428.1 | - | 11 | 1198 | ||
AK098151.1 | - | 6 | 713 | |||
AK223328.1 | - | 10 | 1295 | |||
AK312424.1 | - | 10 | 1305 | |||
BC040239.1 | - | 10 | 1279 | 3047987 | ||
U54617.1 | - | 10 | 1301 |