Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENPP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040092.2 | NP_001035181.1 | 25 | 2664 | ||
NM_001130863.2 | NP_001124335.1 | 26 | 2739 | |||
NM_006209.4 | NP_006200.3 | 26 | 2820 | |||
NR_045555.1 | 14 | 1569 | ||||
XM_006716584.1 | XP_006716647.1 | 27 | 2873 | |||
XM_006716585.1 | XP_006716648.1 | 27 | 2861 | |||
XM_006716587.1 | XP_006716650.1 | 26 | 2786 | |||
XM_017013571.1 | XP_016869060.1 | 26 | 2773 | |||
XM_017013572.1 | XP_016869061.1 | 26 | 2705 | |||
XM_017013573.1 | XP_016869062.1 | 26 | 2761 | |||
XM_017013574.1 | XP_016869063.1 | 25 | 2698 | |||
XM_017013575.1 | XP_016869064.1 | 25 | 2630 | |||
GenBank mRNA | AI375195.1 | - | 1 | 460 | ||
AK124910.1 | - | 7 | 3199 | |||
AK130313.1 | - | 14 | 1569 | |||
AK296027.1 | - | 11 | 1280 | |||
AK315253.1 | - | 25 | 2637 | |||
BC034961.2 | - | 25 | 2637 | 4828484 | ||
D45421.1 | - | 25 | 2611 | |||
EU131011.2 | - | 26 | 2653 | |||
L35594.1 | - | 26 | 2783 |