Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC26A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000441.1 | NP_000432.1 | 17, 18 | 2257 | ||
XM_005250425.2 | XP_005250482.1 | 17, 18 | 2240 | |||
XM_017012318.1 | XP_016867807.1 | 15, 16 | 1958 | |||
GenBank mRNA | AF030880.1 | - | 17, 18 | 2257 | ||
AK294388.1 | - | 8, 9 | 1136 | |||
AK298543.1 | - | 7, 8 | 880 | |||
AK300604.1 | - | 8, 9 | 952 |