Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PENK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135690.2 | NP_001129162.1 | 4 | 921 | ||
GenBank mRNA | AK314908.1 | - | 4 | 862 | ||
BC032505.1 | - | 2 | 772 | 5244387 | ||
BI496610.1 | - | 1 | 441 | |||
CR541808.1 | - | 2 | 696 | |||
CR541828.1 | - | 2 | 696 |