Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEPD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000285.3 | NP_000276.2 | 8, 9 | 758 | ||
NM_001166057.1 | NP_001159529.1 | 6, 7 | 566 | |||
GenBank mRNA | AK225037.1 | - | 8, 9 | 639 | ||
AK290756.1 | - | 8, 9 | 639 | |||
AK290781.1 | - | 8, 9 | 654 | |||
AK291561.1 | - | 8, 9 | 656 | |||
AK293126.1 | - | 6, 7 | 461 | |||
BC004305.1 | - | 8, 9 | 629 | 3626134 | ||
BC015027.1 | - | 8, 9 | 659 | 3920275 | ||
BC028295.1 | - | 8, 9 | 630 | 4470066 | ||
BT006692.1 | - | 8, 9 | 625 | |||
CR541669.1 | - | 8, 9 | 625 | |||
J04605.1 | - | 8, 9 | 641 |