Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEX6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000287.3 | NP_000278.3 | 8 | 1840 | ||
NM_001316313.1 | NP_001303242.1 | 8 | 1576 | |||
NR_133009.1 | 8 | 1840 | ||||
XM_011514661.1 | XP_011512963.1 | 7 | 1756 | |||
GenBank mRNA | AB051076.1 | - | 8 | 1747 | ||
AB051077.1 | - | 8 | 1483 | |||
AB051078.1 | - | 8 | 1747 | |||
AB208906.1 | - | 8 | 1800 | |||
AK225704.1 | - | 7 | 2153 | |||
AK314237.1 | - | 8 | 1776 | |||
BC033487.1 | - | 8 | 1755 | |||
BC048331.1 | - | 8 | 1763 | 5189685 | ||
D83703.1 | - | 8 | 1817 |