Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PFDN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002623.3 | NP_002614.2 | 3 | 314 | ||
XM_017027879.1 | XP_016883368.1 | 3 | 598 | |||
GenBank mRNA | AK312059.1 | - | 3 | 190 | ||
BC010953.1 | - | 3 | 194 | 4273238 | ||
CN482830.1 | - | 3 | 314 | |||
U41816.1 | - | 3 | 175 |