Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PFKP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242339.1 | NP_001229268.1 | 22 | 2285 | ||
NM_001323067.1 | NP_001309996.1 | 20 | 2024 | |||
NM_001323068.1 | NP_001309997.1 | 19 | 2008 | |||
NM_001323069.1 | NP_001309998.1 | 20 | 2156 | |||
NM_001323070.1 | NP_001309999.1 | 14 | 1818 | |||
NM_001323071.1 | NP_001310000.1 | 21 | 2277 | |||
NM_001323072.1 | NP_001310001.1 | 21 | 2384 | |||
NM_001323073.1 | NP_001310002.1 | 16 | 1645 | |||
NM_001323074.1 | NP_001310003.1 | 15 | 1971 | |||
NM_002627.4 | NP_002618.1 | 20 | 2161 | |||
XM_005252465.4 | XP_005252522.1 | 21 | 2359 | |||
XM_005252466.4 | XP_005252523.1 | 20 | 2206 | |||
XM_005252467.2 | XP_005252524.1 | 20 | 2130 | |||
XM_006717449.1 | XP_006717512.1 | 21 | 2283 | |||
GenBank mRNA | AK092597.1 | - | 22 | 2285 | ||
AK097692.1 | - | 4 | 1214 | |||
AK128373.1 | - | 7 | 2673 | |||
AK129843.1 | - | 2017 | ||||
AK130767.1 | - | 5 | 1447 | |||
AK291841.1 | - | 20 | 2133 | |||
AK308003.1 | - | 20 | 2124 | |||
AK309692.1 | - | 13 | 1461 | |||
AL521453.3 | - | 4 | 336 | |||
BC002536.1 | - | 20 | 2121 | 3140233 | ||
BC021698.1 | - | 11 | 1074 | |||
BC029138.1 | - | 20 | 2161 | 5180268 | ||
BC043156.1 | - | 4 | 1857 | |||
BU737488.1 | - | 3 | 506 | |||
CD365858.1 | - | 3 | 506 | |||
D25328.1 | - | 20 | 2118 |
Gene Symbol: PITRM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX649017.1 | - | 26 | 7251 |