Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PFKP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242339.1 | NP_001229268.1 | 10 | 1004 | ||
NM_001323067.1 | NP_001309996.1 | 8 | 743 | |||
NM_001323068.1 | NP_001309997.1 | 8 | 880 | |||
NM_001323069.1 | NP_001309998.1 | 8 | 875 | |||
NM_001323070.1 | NP_001309999.1 | 3 | 690 | |||
NM_001323071.1 | NP_001310000.1 | 9 | 996 | |||
NM_001323072.1 | NP_001310001.1 | 9 | 1103 | |||
NM_001323073.1 | NP_001310002.1 | 4 | 364 | |||
NM_001323074.1 | NP_001310003.1 | 3 | 690 | |||
NM_002627.4 | NP_002618.1 | 8 | 880 | |||
XM_005252465.4 | XP_005252522.1 | 8 | 925 | |||
XM_005252466.4 | XP_005252523.1 | 8 | 925 | |||
XM_005252467.2 | XP_005252524.1 | 8 | 849 | |||
XM_006717449.1 | XP_006717512.1 | 8 | 849 | |||
GenBank mRNA | AK092597.1 | - | 10 | 1004 | ||
AK129843.1 | - | 419 | ||||
AK291841.1 | - | 8 | 852 | |||
AK308226.1 | - | 4 | 280 | |||
BC002536.1 | - | 8 | 840 | 3140233 | ||
BC029138.1 | - | 8 | 880 | 5180268 | ||
D25328.1 | - | 8 | 837 |