Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304451.1 | NP_001291380.1 | 6 | 609 | ||
NM_001304452.1 | NP_001291381.1 | 7 | 754 | |||
NM_002631.3 | NP_002622.2 | 7 | 675 | |||
GenBank mRNA | AK290404.1 | - | 7 | 600 | ||
AK296889.1 | - | 8 | 503 | |||
AK298830.1 | - | 7 | 754 | |||
AK304423.1 | - | 6 | 532 | |||
BC000368.2 | - | 7 | 586 | 2819330 |