Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138733.4 | NP_620061.2 | 1 | 398 | ||
GenBank mRNA | AK312770.1 | - | 1 | 365 | ||
BC038843.1 | - | 1 | 355 | 5741907 | ||
DB043284.1 | - | 1 | 398 | |||
FJ224296.1 | - | 1 | 315 |