Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172818.1 | NP_001166289.1 | 8, 9 | 1574 | ||
NM_001172819.1 | NP_001166290.1 | 8, 9 | 1365 | |||
NM_002633.2 | NP_002624.2 | 8, 9 | 1488 | |||
GenBank mRNA | AK293298.1 | - | 8, 9 | 1365 | ||
AK294186.1 | - | 9, 10 | 1447 | |||
AK298505.1 | - | 8, 9 | 1295 | |||
AK300100.1 | - | 8, 9 | 1560 | |||
AK312254.1 | - | 8, 9 | 1353 | |||
BC001756.2 | - | 8, 9 | 1401 | 3505510 | ||
BC019920.1 | - | 8, 9 | 1488 | 5122261 | ||
BC067763.1 | - | 8, 9 | 1366 | 5286530 | ||
BC090856.1 | - | 8, 9 | 1333 | |||
BT006961.1 | - | 8, 9 | 1275 | |||
M83088.1 | - | 8, 9 | 1337 |