Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172818.1 | NP_001166289.1 | 4 | 924 | ||
NM_001172819.1 | NP_001166290.1 | 4 | 715 | |||
NM_002633.2 | NP_002624.2 | 4 | 838 | |||
GenBank mRNA | AK293298.1 | - | 4 | 715 | ||
AK294186.1 | - | 5 | 797 | |||
AK298505.1 | - | 4 | 645 | |||
AK300100.1 | - | 4 | 910 | |||
AK312254.1 | - | 4 | 703 | |||
BC001756.2 | - | 4 | 751 | 3505510 | ||
BC019920.1 | - | 4 | 838 | 5122261 | ||
BC067763.1 | - | 4 | 716 | 5286530 | ||
BC090856.1 | - | 4 | 683 | |||
BT006961.1 | - | 4 | 625 | |||
M83088.1 | - | 4 | 687 |