Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199917.1 | NP_001186846.1 | 8 | 1167 | ||
NM_001199918.1 | NP_001186847.1 | 6 | 825 | |||
NM_001199919.1 | NP_001186848.1 | 7 | 1031 | |||
NM_015599.2 | NP_056414.1 | 7 | 1031 | |||
XM_017010935.1 | XP_016866424.1 | 6 | 798 | |||
XM_017010936.1 | XP_016866425.1 | 7 | 1030 | |||
XM_017010937.1 | XP_016866426.1 | 6 | 798 | |||
XM_017010938.1 | XP_016866427.1 | 7 | 1019 | |||
GenBank mRNA | AB032081.1 | - | 7 | 943 | ||
AF102265.1 | - | 7 | 947 | |||
AF180371.1 | - | 7 | 954 | |||
AK001365.1 | - | 6 | 766 | |||
AK023432.1 | - | 7 | 976 | |||
AK225363.1 | - | 7 | 969 | |||
AK301867.1 | - | 8 | 1145 | |||
AK314512.1 | - | 7 | 964 | |||
BC001258.1 | - | 7 | 990 | 3451677 |